Bullard™ EVAHL PAPR System For Use with Hazardous Locations
Narrow profile PAPR offers workers protection in hazardous locations. System includes loose-fitting facepiece, blower, belt, battery, breathing tube, charger, air flow indicator, and head top with ratchet suspension.
Supplier:  Bullard™ EVAHL20LFL
 View more versions of this product
Catalog No. 17-989-500
Explore available promotions
Description
Description
  • HazLoc rated†
  • For Class I, Division 2, Groups A, B, C, D T6 (Gases and Vapors)
  • Class II, Division 2, Groups F, G (Dusts)
  • Class III (Fibers or Flyings)
  • Simple, one button operation
  • Choice of two air speeds
  • Single filter design
  • Active Flow Technology responds to air flow conditions
  • 25 Assigned Protection Factor (APF) for Loose Fitting Headtops
  • Independent, 3rd Party Documentation for Assigned Protection Factor of 1000 for certain style Hoods (RT, CC20) and Spectrum Full face mask
  • Dual alarms for low flow and low battery
  • Visual fuel gauge
  • 10X the life of most brush motors
  • Genius Charger compatible
†Not for use in Oxygen deficient atmospheres
Compliance
Meets NIOSH; ANSI™/ISA 2.12.01-2007 and CSA approvals for Class I, Division 2, Groups A, B, C, D, T6; Class II, Division 2 Groups F, G; Class III.
†CSA International Certificate 2510928 per ANSI/ISA 12.12.01-2007
Specifications
Specifications
ANSI/ISA 2.12.01-2007
Lithium-ion Polymer
NIOSH
PAPR System with Facepiece
Painting, chemical handling, healthcare, pharmaceutical
7 to 8 hours
Foam
CSA Class I, Division 2, Groups A, B, C, D, T6, Class II, Division 2 Groups F, G, Class III
8.5/7 CFM
EVAHL20LFL