Bullard™ EVAHL PAPR System For Use with Hazardous Locations
Narrow profile PAPR offers workers protection in hazardous locations. System includes loose-fitting facepiece, blower, belt, battery, breathing tube, charger, air flow indicator, and head top with ratchet suspension.
Supplier: Bullard™ EVAHL20LFM
Description
- HazLoc rated†
- For Class I, Division 2, Groups A, B, C, D T6 (Gases and Vapors)
- Class II, Division 2, Groups F, G (Dusts)
- Class III (Fibers or Flyings)
- Simple, one button operation
- Choice of two air speeds
- Single filter design
- Active Flow Technology responds to air flow conditions
- 25 Assigned Protection Factor (APF) for Loose Fitting Headtops
- Independent, 3rd Party Documentation for Assigned Protection Factor of 1000 for certain style Hoods (RT, CC20) and Spectrum Full face mask
- Dual alarms for low flow and low battery
- Visual fuel gauge
- 10X the life of most brush motors
- Genius Charger compatible
Compliance
Meets NIOSH; ANSI™/ISA 2.12.01-2007 and CSA approvals for Class I, Division 2, Groups A, B, C, D, T6; Class II, Division 2 Groups F, G; Class III.
†CSA International Certificate 2510928 per ANSI/ISA 12.12.01-2007
Specifications
ANSI/ISA 2.12.01-2007 | |
Lithium-ion Polymer | |
NIOSH | |
PAPR System with Facepiece | |
8.5/7 CFM | |
EVAHL20LFM |
7 to 8 hours | |
Foam | |
CSA Class I, Division 2, Groups A, B, C, D, T6, Class II, Division 2 Groups F, G, Class III | |
Medium | |
Painting, chemical handling, healthcare, pharmaceutical |