Bullard™ Evolutionary Air Hazardous Location (EVAHL) PAPR System Accessory, Double Bib Hood
Designed to be used with the EVAHL Powered Air Purifying Respirator
Supplier:  Bullard™ EVAHLRT3TDB
 View more versions of this product
Catalog No. 17-989-531
Explore available promotions
Description
Description
  • Ideal for painting, chemical handling, healthcare, pharmaceutical applications
  • Choice of hood configurations
  • Airflow rate: 8.5/7 cfm
  • 77dBA low voltage and 77dBA low flow alarm
  • Lithium ion polymer battery (1-year life span)
Certifications
ISO 9001, NIOSH, CSA, Class I Division 2, Class II Division 2, Class III
Specifications
Specifications
Lithium Ion Polymer
ISO 9001, NIOSH, CSA, Class I Division 2, Class II Division 2, Class III
Double Bib Suspension-free Hood with Vinyl Decon Belt
Ideal for painting, chemical handling, healthcare, pharmaceutical