Bullard™ Loose Fitting Facepieces for PA20, PA30, PA30IS, EVA, EVAHL PAPRs
Loose fitting facepieces provide comfortable and effective protection from a wide range of respiratory hazards.
Supplier: Bullard™ 20LF2S
Description
- Loose fitting facepiece
- Head covers made of Tychem
- Sewn-in headband suspension
- Twist-lock breathing tube connection
- For use with PAPR and SAR systems
- Choose from a variety of sizes and color connections
- Acetate lens material
- NIOSH approved
Specifications
25 | |
Small | |
Pa20, pa30, pa30is, eva™, eva™hl powered air purifying respirator | |
Yes |
Loose Fitting Facepiece | |
Pharmaceutical manufacturing, general industry and maintenance, woodworking, spray foam, painting, healthcare | |
Narrow profile, green twist lock connection | |
20LF2S |