Bullard™ Loose Fitting Facepieces for PA20, PA30, PA30IS, EVA, EVAHL PAPRs
Loose fitting facepieces provide comfortable and effective protection from a wide range of respiratory hazards.
Supplier:  Bullard™ 20LF2L
 View more versions of this product
Catalog No. 29-635-9915
Explore available promotions
Description
Description
  • Loose fitting facepiece
  • Head covers made of Tychem
  • Sewn-in headband suspension
  • Twist-lock breathing tube connection
  • For use with PAPR and SAR systems
  • Choose from a variety of sizes and color connections
  • Acetate lens material
  • NIOSH approved
Specifications
Specifications
25
Large
Pa20, pa30, pa30is, eva™, eva™hl powered air purifying respirator
20LF2L
Loose Fitting Facepiece
Pharmaceutical manufacturing, general industry and maintenance, woodworking, spray foam, painting, healthcare
Narrow profile, black quick-attach twist lock connection