Bullard™ RT Series Tychem™ Hood Respirator Assemblies
Delivers compressed air through certified breathing air hose to workers wearing facemasks
Supplier:  Bullard™ RT130
 View more versions of this product
Catalog No. 36-100-3796
Explore available promotions
Description
Description
RT Series Hood can be used for both Powered Air-Purifying Respirators (PAPR) and Supplied Air Respirators (SAR).
  • Headband-free design
  • Choice of Tychem QC or SL
  • 1000 Assigned Protection Factor (APF)
  • Panoramic viewing lens
  • Sport neck cuff
  • Long, double bibs
  • Taped and sealed seams (RT4 only)
  • No fit testing needed
The RT Series Hoods are available in several configurations to meet most levels of disposability requirements. As with most Tychem hoods, the RT Series is available as a stand-alone replacement hood for those applications that allow for decontamination and reuse of the breathing tube. For single use applications, the RT Series is available fully assembled with a disposable lightweight breathing tube. Individually packaged with the breathing tube already attached, the user needs only to don the hood and connect the flow control device. After exiting from the area, the user may dispose of the complete assembly.
Specifications
Specifications
Tychem QC
Supplied air respirator
RT130
Supplied Air Respirator Assembly
RT1 hood, 20BT breathing tube, F30 flow control