Bullard™ Spectrum™ Series Full-Facepiece Airline Respirators
Continuous-flow respirators feature lightweight facepieces with a wide-angle lens for a clear field of view.
Supplier:  Bullard™ SPEC40L
 View more versions of this product
Catalog No. 29-663-318
Explore available promotions
Description
Description
  • Continuous-flow, high-pressure model includes breathing tube and flow valve for compressor use
  • Continuous-flow, low-pressure model includes breathing tube and constant flow valve for air pump use
  • Silicone flange seals for comfortable fit
  • SnapFit™ design for easy donning
  • Clear silicone nosecup and low-profile regulator minimize interference with vision and other PPE
  • Regulator allows donning without free flow of air, conserving air supply
  • Swivel connector provides free movement of regulator hose for comfort and maneuverability
  • All models equipped with medium/large facepiece
  • Can be used with V10 and V20 Series breathing air hoses (order separately)
Compliance
NIOSH approved.
Specifications
Specifications
NIOSH
Medium/Large
Headstrap, breathing tube assembly with flow control device and nylon belt
High Pressure Respirator Assembly
Breathing air compressor
SPEC40L