Bulwark™ FR Excel FR™ Arc-Rated Coveralls
Exceptional fit and protection
Supplier:  Bulwark™ FR CED2NVLN44
 View more versions of this product
Catalog No. 19-602-011
Explore available promotions
Description
Description
  • One-piece coverall made from flame-resistant 9 oz. Twill 100% cotton
  • With arc rating ATPV 10.6 calories/cm2
  • Two-way concealed Nomex™ taped brass break-away zipper with hidden snap at top
  • Topstitched, lay-flat collar
  • Deep-pleated action back and side vents with snap closures
  • Elastic waist inserts ensure comfortable fit
  • Sleeve cuffs with concealed snap closure
  • Features eight pockets: two lined and topstitched front swing pockets; two hip pockets with hidden snap closures; two chest pockets with flaps and concealed snaps; one rule pocket on right leg; and one pocket on left sleeve
Specifications
Specifications
Men's
Navy
Collared
Flame
11.2 Calories/cm2
8
42.5 to 44 in.
113 cm
1.55 to 1.92 m
Cotton
Elastic
2-way Zipper
Lay flat
NFPA 2112
44 in.
44.5 in.
5.11 to 6.3 ft.
Long