Learn More
Bupivacaine hydrochloride monohydrate, 98+%, Thermo Scientific Chemicals
Inhibits TREK-1 channels and depolarizes the cell membrane
$121.07 - $1694.70
Chemical Identifiers
CAS | 73360-54-0 |
---|---|
Molecular Formula | C18H31ClN2O2 |
Molecular Weight (g/mol) | 342.908 |
MDL Number | MFCD00078956 |
InChI Key | HUCIWBPMHXGLFM-UHFFFAOYSA-N |
Synonym | bupivacaine hydrochloride monohydrate, lac-43, marcaine tn, +/--1-butyl-2',6'-pipecoloxylidide monohydrochloride, monohydrate, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide hydrate hydrochloride, bupivacaine hydrate hydrochloride, bupivacaine hcl h2-o, 2-piperidinecarboxamide, 1-butyl-n-2,6-dimethylphenyl-, monohydrochloride, monohydrate, bupivacaine hydrochloride ep, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide hydrochloride hydrate |
PubChem CID | 5282419 |
IUPAC Name | 1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide;hydrate;hydrochloride |
SMILES | CCCCN1CCCCC1C(=O)NC2=C(C=CC=C2C)C.O.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6283503
|
Thermo Scientific Chemicals
J6283503 |
1 g |
Each for $121.07
|
|
|||||
AAJ6283506
|
Thermo Scientific Chemicals
J6283506 |
5 g |
Each for $388.97
|
|
|||||
AAJ6283514
|
Thermo Scientific Chemicals
J6283514 |
25 g |
Each for $1,694.70
|
|
|||||
Description
It is employed as Na+ channel blocker, local anesthetic and cAMP production inhibitor. It acts as a surfactant molecule possessing both hydrophilic and lipophilic properties, adrenergic antagonist, EC 3.1.1.8 (cholinesterase) inhibitor and EC 3.6.3.8 (Ca(2+)-transporting ATPase) inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as Na+ channel blocker, local anesthetic and cAMP production inhibitor. It acts as a surfactant molecule possessing both hydrophilic and lipophilic properties, adrenergic antagonist, EC 3.1.1.8 (cholinesterase) inhibitor and EC 3.6.3.8 (Ca(2+)-transporting ATPase) inhibitor.
Solubility
Soluble in water to 50 mM, in DMSO to 100 mM and in ethanol to 100 mM.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
73360-54-0 | |
342.908 | |
HUCIWBPMHXGLFM-UHFFFAOYSA-N | |
5282419 | |
CCCCN1CCCCC1C(=O)NC2=C(C=CC=C2C)C.O.Cl |
C18H31ClN2O2 | |
MFCD00078956 | |
bupivacaine hydrochloride monohydrate, lac-43, marcaine tn, +/--1-butyl-2',6'-pipecoloxylidide monohydrochloride, monohydrate, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide hydrate hydrochloride, bupivacaine hydrate hydrochloride, bupivacaine hcl h2-o, 2-piperidinecarboxamide, 1-butyl-n-2,6-dimethylphenyl-, monohydrochloride, monohydrate, bupivacaine hydrochloride ep, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide hydrochloride hydrate | |
1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide;hydrate;hydrochloride |
Specifications
73360-54-0 | |
White | |
MFCD00078956 | |
UN2811 | |
bupivacaine hydrochloride monohydrate, lac-43, marcaine tn, +/--1-butyl-2',6'-pipecoloxylidide monohydrochloride, monohydrate, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide hydrate hydrochloride, bupivacaine hydrate hydrochloride, bupivacaine hcl h2-o, 2-piperidinecarboxamide, 1-butyl-n-2,6-dimethylphenyl-, monohydrochloride, monohydrate, bupivacaine hydrochloride ep, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide hydrochloride hydrate | |
HUCIWBPMHXGLFM-UHFFFAOYSA-N | |
1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide;hydrate;hydrochloride | |
5282419 | |
≥98% | |
Bupivacaine hydrochloride monohydrate |
255°C to 259°C | |
C18H31ClN2O2 | |
1 g | |
14,1495 | |
Soluble in water to 50 mM,in DMSO to 100 mM and in ethanol to 100 mM. | |
CCCCN1CCCCC1C(=O)NC2=C(C=CC=C2C)C.O.Cl | |
342.908 | |
342.9 | |
Powder |
Safety and Handling
GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P330-P361-P363-P501c
H300+H310+H330
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 241-917-8
RTECSNumber : TK6125000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only