Learn More
Thermo Scientific Chemicals Burgess Reagent, 96%
Supplier: Thermo Scientific Chemicals L2015503
Description
Burgess reagent is a selective dehydrating reagent used in organic synthesis. It is used to convert secondary and tertiary alcohol, with an adjacent proton, into alkenes. Burgess reagent is also utilized to promote great synthetic values in medicinal chemistry. It has been used in dehydration of primary amides, formamides and primary nitroalkanes to generate nitriles, isocyanides and nitrile oxides respectively. It forms urethanes when it reacts with primary alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBurgess reagent is a selective dehydrating reagent used in organic synthesis. It is used to convert secondary and tertiary alcohol, with an adjacent proton, into alkenes. Burgess reagent is also utilized to promote great synthetic values in medicinal chemistry. It has been used in dehydration of primary amides, formamides and primary nitroalkanes to generate nitriles, isocyanides and nitrile oxides respectively. It forms urethanes when it reacts with primary alcohols.
Solubility
Soluble in organic solvents.
Notes
Store in cool place. Moisture sensitive. Incompatible with strong oxidizing agents.
Specifications
Burgess Reagent | |
72°C to 82°C | |
MFCD00077815 | |
1432131 | |
methoxycarbonylsulfamoyl triethylammonium hydroxide, burgess reagent, carbomethoxysulfamoyltriethylammonium hydroxide, methoxycarbonylsulfamoyltriethylammonium hydroxide, methoxycarbonylsulphamoyltriethylammonium hydroxide, methoxy-carbonylsulfamoyltriethylammonium hydroxide, methoxycarbonylsulfa-moyl-triethylammonium hydroxide, methoxycarbonylsulphamoyl-triethylammonium hydroxide | |
YSHOWEKUVWPFNR-UHFFFAOYSA-N | |
(methoxycarbonyl)[(triethylazaniumyl)sulfonyl]azanide | |
11032497 | |
96% |
29684-56-8 | |
C8H18N2O4S | |
1 g | |
Moisture sensitive | |
Soluble in organic solvents. | |
CC[N+](CC)(CC)S(=O)(=O)[N-]C(=O)OC | |
238.30 | |
238.3 |
Chemical Identifiers
29684-56-8 | |
238.30 | |
YSHOWEKUVWPFNR-UHFFFAOYSA-N | |
11032497 |
C8H18N2O4S | |
MFCD00077815 | |
methoxycarbonylsulfamoyl triethylammonium hydroxide, burgess reagent, carbomethoxysulfamoyltriethylammonium hydroxide, methoxycarbonylsulfamoyltriethylammonium hydroxide, methoxycarbonylsulphamoyltriethylammonium hydroxide, methoxy-carbonylsulfamoyltriethylammonium hydroxide, methoxycarbonylsulfa-moyl-triethylammonium hydroxide, methoxycarbonylsulphamoyl-triethylammonium hydroxide | |
CC[N+](CC)(CC)S(=O)(=O)[N-]C(=O)OC |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only