Learn More
Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
$111.34 - $183.46
Chemical Identifiers
CAS | 94-26-8 |
---|---|
Molecular Formula | C11H14O3 |
Molecular Weight (g/mol) | 194.23 |
MDL Number | MFCD00016478 |
InChI Key | QFOHBWFCKVYLES-UHFFFAOYSA-N |
Synonym | butylparaben, butyl paraben, butyl p-hydroxybenzoate, nipabutyl, butoben, butyl chemosept, butyl parasept, butyl tegosept, butyl butex, tegosept butyl |
PubChem CID | 7184 |
IUPAC Name | butyl 4-hydroxybenzoate |
SMILES | CCCCOC(=O)C1=CC=C(C=C1)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC403571000
|
Thermo Scientific Chemicals
403571000 |
100 g | Glass bottle |
Each for $111.34
|
|
||||
AC403575000
|
Thermo Scientific Chemicals
403575000 |
500 g | Glass bottle |
Each for $183.46
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
94-26-8 | |
194.23 | |
QFOHBWFCKVYLES-UHFFFAOYSA-N | |
7184 | |
CCCCOC(=O)C1=CC=C(C=C1)O |
C11H14O3 | |
MFCD00016478 | |
butylparaben, butyl paraben, butyl p-hydroxybenzoate, nipabutyl, butoben, butyl chemosept, butyl parasept, butyl tegosept, butyl butex, tegosept butyl | |
butyl 4-hydroxybenzoate |
Specifications
94-26-8 | |
White | |
156.0°C (3.5 mmHg) | |
Authentic | |
Glass bottle | |
OHC6H4CO2(CH2)3CH3 | |
100 g | |
butylparaben, butyl paraben, butyl p-hydroxybenzoate, nipabutyl, butoben, butyl chemosept, butyl parasept, butyl tegosept, butyl butex, tegosept butyl | |
QFOHBWFCKVYLES-UHFFFAOYSA-N | |
butyl 4-hydroxybenzoate | |
7184 | |
99+% | |
Butyl 4-hydroxybenzoate, 99+% |
68.0°C to 71.0°C | |
1.2800g/mL | |
181°C | |
99% min. (GC) | |
C11H14O3 | |
MFCD00016478 | |
15, 1587 | |
Solubility in water: slightly soluble (1.8wt% at 20°C). Other solubilities: soluble in acetone,ethanol,diethyl ether,,chloroform,and propylene glycol | |
CCCCOC(=O)C1=CC=C(C=C1)O | |
194.23 | |
194.23 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
EINECSNumber : 202-318-7
RTECSNumber : DH1980000
TSCA : TSCA
RUO – Research Use Only