missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Butyl Acetoacetate 98.0+%, TCI America™
Supplier: TCI America A079525ML
Specifications
Butyl Acetoacetate | |
Colorless | |
C8H14O3 | |
25 mL | |
REIYHFWZISXFKU-UHFFFAOYSA-N | |
butyl 3-oxobutanoate | |
11576 | |
≥98.0% (GC) |
591-60-6 | |
214°C | |
MFCD00059252 | |
Acetoacetic Acid Butyl Ester | |
CCCCOC(=O)CC(C)=O | |
158.20 | |
158.20 | |
Liquid |
Chemical Identifiers
591-60-6 | |
158.20 | |
REIYHFWZISXFKU-UHFFFAOYSA-N | |
11576 | |
CCCCOC(=O)CC(C)=O |
C8H14O3 | |
MFCD00059252 | |
Acetoacetic Acid Butyl Ester | |
butyl 3-oxobutanoate |
Safety and Handling
EINECSNumber : (2)-1475&(2)-1505
RTECSNumber : AK5100000
TSCA : Yes