missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Butyl acrylate, 99+%, stabilized, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 215860100

 View more versions of this product

Catalog No. AC215860100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Butyl acrylate
0.01% max. (as acrylic acid)
0.8900g/mL
39°C
99% min. (GC)
C7H12O2
H2C=CHCO2(CH2)3CH3
10 L
0.89
butyl acrylate, n-butyl acrylate, acrylic acid butyl ester, n-butyl propenoate, 2-propenoic acid, butyl ester, butyl 2-propenoate, butylacrylate, acrylic acid, butyl ester, acrylic acid n-butyl ester, butylester kyseliny akrylove
CQEYYJKEWSMYFG-UHFFFAOYSA-N
butyl prop-2-enoate
10 to 20ppm MEHQ
0.869 mPa.s (20°C)
128.17
Liquid
141-32-2,150-76-5
-64.0°C
145.0°C
Authentic
Plastic drum
1.4170 to 1.4190
MFCD00009446
02, II, 388
15, 1541
Solubility in water: 1.4g/L (20°C)
CCCCOC(=O)C=C
128.17
8846
CHEBI:3245
99+%

Chemical Identifiers

141-32-2
128.17
CQEYYJKEWSMYFG-UHFFFAOYSA-N
8846
butyl prop-2-enoate
C7H12O2
MFCD00009446
butyl acrylate, n-butyl acrylate, acrylic acid butyl ester, n-butyl propenoate, 2-propenoic acid, butyl ester, butyl 2-propenoate, butylacrylate, acrylic acid, butyl ester, acrylic acid n-butyl ester, butylester kyseliny akrylove
CHEBI:3245
CCCCOC(=O)C=C
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful to aquatic li

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to f

GHS Signal Word: Warning

EINECSNumber : 205-480-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only