missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyl Carbamate 98.0+%, TCI America™
$113.71 - $993.54
Chemical Identifiers
| CAS | 592-35-8 |
|---|---|
| Molecular Formula | C5H11NO2 |
| Molecular Weight (g/mol) | 117.148 |
| MDL Number | MFCD00007967 |
| InChI Key | SKKTUOZKZKCGTB-UHFFFAOYSA-N |
| Synonym | n-butyl carbamate, carbamic acid, butyl ester, usaf fo-1, usaf el-101, carbamic acid butyl ester, unii-my8i82ajs5, ccris 5069, my8i82ajs5, butyl aminooate, o-butyl carbamate |
| PubChem CID | 11596 |
| IUPAC Name | butyl carbamate |
| SMILES | CCCCOC(=O)N |
Chemical Identifiers
| 592-35-8 | |
| 117.148 | |
| SKKTUOZKZKCGTB-UHFFFAOYSA-N | |
| 11596 | |
| CCCCOC(=O)N |
| C5H11NO2 | |
| MFCD00007967 | |
| n-butyl carbamate, carbamic acid, butyl ester, usaf fo-1, usaf el-101, carbamic acid butyl ester, unii-my8i82ajs5, ccris 5069, my8i82ajs5, butyl aminooate, o-butyl carbamate | |
| butyl carbamate |
Specifications
| 54°C | |
| 204°C | |
| C5H11NO2 | |
| n-butyl carbamate, carbamic acid, butyl ester, usaf fo-1, usaf el-101, carbamic acid butyl ester, unii-my8i82ajs5, ccris 5069, my8i82ajs5, butyl aminooate, o-butyl carbamate | |
| CCCCOC(=O)N | |
| 25 g | |
| 11596 | |
| ≥98.0% (GC) | |
| Butyl Carbamate |
| 592-35-8 | |
| White | |
| MFCD00007967 | |
| SKKTUOZKZKCGTB-UHFFFAOYSA-N | |
| butyl carbamate | |
| 117.148 | |
| 117.15 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-1712
RTECSNumber : EZ0175000
TSCA : Yes