missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Butyl Formate 95.0+%, TCI America™

Supplier:  TCI America F005125ML

 View more versions of this product

Catalog No. F005125ML


Explore available promotions

Description

Description

Specifications

Specifications

Butyl Formate
-91°C
107°C
MFCD00003296
1128
NMJJFJNHVMGPGM-UHFFFAOYSA-N
butyl formate
11614
≥95.0% (GC)
592-84-7
Yellow
C5H10O2
25 mL
n-butyl formate, formic acid, butyl ester, butyl methanoate, n-butyl methanoate, formic acid butyl ester, n-butylformate, butylester kyseliny mravenci, fema no. 2196, unii-0y9mz7vm9p, butylester kyseliny mravenci czech
CCCCOC=O
102.13
102.13
Liquid

Chemical Identifiers

592-84-7
102.13
NMJJFJNHVMGPGM-UHFFFAOYSA-N
11614
CCCCOC=O
C5H10O2
MFCD00003296
n-butyl formate, formic acid, butyl ester, butyl methanoate, n-butyl methanoate, formic acid butyl ester, n-butylformate, butylester kyseliny mravenci, fema no. 2196, unii-0y9mz7vm9p, butylester kyseliny mravenci czech
butyl formate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0684

RTECSNumber : LQ5500000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions