missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Butyl Methyl Ether 99.0+%, TCI America™
Supplier: TCI America M0504500ML
Specifications
Butyl Methyl Ether | |
Colorless | |
C5H12O | |
500 mL | |
1-Methoxybutane | |
CCCCOC | |
88.15 | |
88.15 | |
Liquid |
628-28-4 | |
70°C | |
MFCD00009453 | |
2350 | |
CXBDYQVECUFKRK-UHFFFAOYSA-N | |
1-methoxybutane | |
12338 | |
≥99.0% (GC) |
Chemical Identifiers
628-28-4 | |
88.15 | |
CXBDYQVECUFKRK-UHFFFAOYSA-N | |
12338 | |
CCCCOC |
C5H12O | |
MFCD00009453 | |
1-Methoxybutane | |
1-methoxybutane |
Safety and Handling
RTECSNumber : KN5240000
TSCA : No