missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva Butyl Sepharose™ 4 Fast Flow Chromatography Media
Used for capture and intermediate purification of larger proteins
Supplier: Cytiva 17098001
Description
- Based on Sepharose Fast Flow base matrix derivatized via uncharged, chemically-stable ether linkages
- Hydrophobic interaction chromatography (HIC) media composed of cross-linked 4% agarose
- Aliphatic butyl ligand immobilized to base matrix with ether linkage
- Fast Flow matrix gives the adsorbent high chemical, physical and thermal stabilities
- Dynamic capacity at low salt concentration
- Used with all aqueous buffers commonly used in chromatography
- Autoclavable at 121°C for 20 minutes in water
- Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development
Specifications
200 mL | |
Stable in commonly used aqueous Buffers: 1 M NaOH, 30% Isopropanol, 70% Ethanol, 6 M Guanidine-Hydrochloride, 30% Acetonitrile, 1 mM HCl | |
Butyl Sepharose 4 Fast Flow Media | |
45 to 165 μm | |
25 cm | |
Based on cross-linked 4% agarose | |
20% Ethanol |
For Capture and Intermediate PurifiCatIon of Larger Proteins | |
4°C to 30°C | |
150 to 250 cm/Hoμr | |
9.84 in. | |
100 kPa | |
3 to 13 | |
Butyl |