missing translation for 'onlineSavingsMsg'
Learn More

Butylated Hydroxytoluene, Granular, NF, 99-101.5%, Spectrum™ Chemical

B1196, 128-37-0, C15H24O

$206.00 - $5904.46

Chemical Identifiers

CAS 128-37-0
Molecular Formula C15H24O
Molecular Weight (g/mol) 220.36
InChI Key NLZUEZXRPGMBCV-UHFFFAOYSA-N
IUPAC Name 2,6-di-tert-butyl-4-methylphenol
SMILES CC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18601166
View Documents
Spectrum Chemical Mfg Cor
B1196125GM
125 g Fiber Drum
Each for $206.00
Only null left
 
18601167
View Documents
Spectrum Chemical Mfg Cor
B1196500GM
500 g Fiber Drum
Each for $512.57
Only null left
 
18601168
View Documents
Spectrum Chemical Mfg Cor
B119625KG
2.5 kg Fiber Drum
Each for $1,595.11
Only null left
 
18601170
View Documents
Spectrum Chemical Mfg Cor
B119612KGBL
12 kg Fiber Drum
Each for $5,904.46
Only null left
 
Description

Description

Spectrum™ Chemical Butylated Hydroxytoluene, Granular, NF is used for its antioxidant properties in pharmaceutical gels, creams, liquids, gelatin capsules and tablets. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

128-37-0
220.36
2,6-di-tert-butyl-4-methylphenol
C15H24O
NLZUEZXRPGMBCV-UHFFFAOYSA-N
CC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C

Specifications

128-37-0
0.002%
C15H24O
NLZUEZXRPGMBCV-UHFFFAOYSA-N
2,6-di-tert-butyl-4-methylphenol
99 to 101.5%
Crystalline Powder or Lumps
100%
Fiber Drum
125 g
CC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C
220.36
NF
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'