missing translation for 'onlineSavingsMsg'
Learn More

Butyraldehyde, 99%

CAS: 123-72-8 | C4H8O | 72.11 g/mol

$82.19 - $206.57

Chemical Identifiers

CAS 123-72-8
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00007023
InChI Key ZTQSAGDEMFDKMZ-UHFFFAOYSA-N
Synonym butyraldehyde, n-butyraldehyde, butyral, 1-butanal, butyric aldehyde, butanaldehyde, butaldehyde, butalyde, butal, n-butanal
PubChem CID 261
ChEBI CHEBI:15743
IUPAC Name butanal
SMILES CCCC=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC108091000
SDS View Documents
Thermo Scientific Chemicals
108091000
100 mL Glass bottle
Each for $82.19
Only null left
 
AC108090010
SDS View Documents
Thermo Scientific Chemicals
108090010
1 L Glass bottle
Each for $105.98
Only null left
 
AC108090025
SDS View Documents
Thermo Scientific Chemicals
108090025
2.5 L Glass bottle
Each for $206.57
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

123-72-8
72.11
ZTQSAGDEMFDKMZ-UHFFFAOYSA-N
261
butanal
C4H8O
MFCD00007023
butyraldehyde, n-butyraldehyde, butyral, 1-butanal, butyric aldehyde, butanaldehyde, butaldehyde, butalyde, butal, n-butanal
CHEBI:15743
CCCC=O

Specifications

123-72-8
Colorless
75.0°C
Authentic
Glass bottle
1.3770 to 1.3830
MFCD00007023
01, 662
15, 1595
Solubility in water: 7.1% (25°C). Other solubilities: miscible with ethanol, ether, ethyl acetate,, acetone and toluene
CCCC=O
72.11
0.43 mPa.s (20°C)
72.11
Liquid
-96.0°C
0.8170g/mL
−12°C
98.5% min. (GC)
C4H8O
CH3CH2CH2CHO
100 mL
0.817
butyraldehyde, n-butyraldehyde, butyral, 1-butanal, butyric aldehyde, butanaldehyde, butaldehyde, butalyde, butal, n-butanal
ZTQSAGDEMFDKMZ-UHFFFAOYSA-N
butanal
261
CHEBI:15743
99%
Butyraldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.

GHS Signal Word: Danger

EINECSNumber : 204-646-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only