missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Butyraldehyde 98.0+%, TCI America™
$39.51 - $55.30
Chemical Identifiers
CAS | 123-72-8 |
---|---|
Molecular Formula | C4H8O |
Molecular Weight (g/mol) | 72.11 |
MDL Number | MFCD00007023 |
InChI Key | ZTQSAGDEMFDKMZ-UHFFFAOYSA-N |
Synonym | butyraldehyde, n-butyraldehyde, butyral, 1-butanal, butyric aldehyde, butanaldehyde, butaldehyde, butalyde, butal, n-butanal |
PubChem CID | 261 |
ChEBI | CHEBI:15743 |
IUPAC Name | butanal |
SMILES | CCCC=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
B075125ML
|
TCI America
B075125ML |
25 mL |
Each for $39.51
|
|
||||||
B0751500ML
|
TCI America
B0751500ML |
500 mL |
Each for $55.30
|
|
||||||
Chemical Identifiers
123-72-8 | |
72.11 | |
ZTQSAGDEMFDKMZ-UHFFFAOYSA-N | |
261 | |
butanal |
C4H8O | |
MFCD00007023 | |
butyraldehyde, n-butyraldehyde, butyral, 1-butanal, butyric aldehyde, butanaldehyde, butaldehyde, butalyde, butal, n-butanal | |
CHEBI:15743 | |
CCCC=O |
Specifications
123-72-8 | |
75°C | |
MFCD00007023 | |
1129 | |
ZTQSAGDEMFDKMZ-UHFFFAOYSA-N | |
butanal | |
261 | |
72.11 | |
Liquid |
Yellow | |
C4H8O | |
25 mL | |
butyraldehyde, n-butyraldehyde, butyral, 1-butanal, butyric aldehyde, butanaldehyde, butaldehyde, butalyde, butal, n-butanal | |
CCCC=O | |
72.11 | |
CHEBI:15743 | |
≥98.0% (GC) | |
Butyraldehyde |
Safety and Handling
EINECSNumber : (2)-0494
RTECSNumber : ES2275000
TSCA : Yes