missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Butyramide, 98%, Thermo Scientific Chemicals

$87.66 - $577.07

Chemical Identifiers

CAS 541-35-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.122
MDL Number MFCD00041894
InChI Key DNSISZSEWVHGLH-UHFFFAOYSA-N
Synonym butyramide, n-butyramide, n-butylamide, butanimidic acid, butanoic acid, amide, unii-9j6or937vr, butyroamide, n-c3h7c o nh2, butyrylamide, butyramid
PubChem CID 10927
ChEBI CHEBI:50724
IUPAC Name butanamide
SMILES CCCC(=O)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1249418
View Documents
Thermo Scientific Chemicals
A1249418
50 g
Each for $87.66
 
AAA1249422
View Documents
Thermo Scientific Chemicals
A1249422
100 g
Each for $147.38
 
AAA1249436
View Documents
Thermo Scientific Chemicals
A1249436
500 g
Each for $577.07
 
Description

Description

Butyramide was used in the synthesis of hydroxamic acids, electrorheological fluids and β-amodoorganotin compounds. It was used as substrate of (+)-γ-lactamase to develop a microreactor to study enzyme stability, activity, kinetics and substrate specificity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Butyramide was used in the synthesis of hydroxamic acids, electrorheological fluids and β-amodoorganotin compounds. It was used as substrate of (+)-γ-lactamase to develop a microreactor to study enzyme stability, activity, kinetics and substrate specificity.

Solubility
Soluble in water (170 g/L).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

541-35-5
87.122
DNSISZSEWVHGLH-UHFFFAOYSA-N
10927
butanamide
C4H9NO
MFCD00041894
butyramide, n-butyramide, n-butylamide, butanimidic acid, butanoic acid, amide, unii-9j6or937vr, butyroamide, n-c3h7c o nh2, butyrylamide, butyramid
CHEBI:50724
CCCC(=O)N

Specifications

541-35-5
1.032
C4H9NO
50 g
14,1592
Soluble in water (170g/L).
CCCC(=O)N
87.122
CHEBI:50724
98%
113°C to 116°C
216°C
MFCD00041894
1361528
butyramide, n-butyramide, n-butylamide, butanimidic acid, butanoic acid, amide, unii-9j6or937vr, butyroamide, n-c3h7c o nh2, butyrylamide, butyramid
DNSISZSEWVHGLH-UHFFFAOYSA-N
butanamide
10927
87.12
Butyramide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 208-776-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only