missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Butyryl chloride, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 149210010

Catalog No. AC149210010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Butyryl chloride
1.0180g/mL
18°C
99%
C4H7ClO
CH3CH2CH2COCl
1L
1.018
butyryl chloride, n-butyryl chloride, butyric acid chloride, butyroyl chloride, butyric chloride, butyrylchloride, unii-2xvm8e16ir, ccris 5989, n-butyric acid chloride, n-butanoyl chloride
DVECBJCOGJRVPX-UHFFFAOYSA-N
butanoyl chloride
8855
99%
141-75-3
102.0°C
Authentic
Glass bottle
1.4112 to 1.4132
MFCD00000752
02, 274
15, 1602
Solubility in water: decomposes. Other solubilities: miscible with ether,decomposes in alcohol
CCCC(=O)Cl
106.55
106.55
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Toxic if inhaled.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 205-498-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only