missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cacodylic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 318150100

 View more versions of this product

Catalog No. AC318150100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cacodylic acid
75-60-5
98%
(CH3)2As(O)OH
15, 1609
OGGXGZAMXPVRFZ-UHFFFAOYSA-N
dimethylarsinic acid
2513
138.01
10 g
Glass bottle
195°C to 198°C
White
C2H7AsO2
MFCD00002095
cacodylic acid, hydroxydimethylarsine oxide, chexmate, silvisar, phytar, arsan, dimethylarsenic acid, erase, arsinic acid, dimethyl, silvisar 510
C[As](C)(O)=O
138.00
CHEBI:48765
≥97.5%
Authentic
Crystalline Powder

Chemical Identifiers

75-60-5
138.00
OGGXGZAMXPVRFZ-UHFFFAOYSA-N
2513
dimethylarsinic acid
C2H7AsO2
MFCD00002095
cacodylic acid, hydroxydimethylarsine oxide, chexmate, silvisar, phytar, arsan, dimethylarsenic acid, erase, arsinic acid, dimethyl, silvisar 510
CHEBI:48765
C[As](C)(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only