Cadence Science™ Flexible Plastic (PTFE) Needles with Bite Protectors
Covered at the hub end with 304 stainless steel tubing – protects tubing from damage due to biting during use.
Supplier: Cadence Science™ POP9926B
Specifications
Single-use | |
17G | |
3.3 in. |
Animal feeding | |
Sterile | |
2.4 mm |