missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cadmium carbonate, 97%, Thermo Scientific Chemicals

$53.73 - $438.27

Chemical Identifiers

CAS 513-78-0
Molecular Formula CCdO3
Molecular Weight (g/mol) 172.42
MDL Number MFCD00010918
InChI Key GKDXQAKPHKQZSC-UHFFFAOYSA-L
Synonym cadmium carbonate, mikrokalcit, otavite, kalcit, supermikrokalcit, carbonic acid, cadmium salt 1:1, cadmium monocarbonate, chemcarb cdco3, caswell no. 134a, unii-616zvs3bly
PubChem CID 10564
SMILES [Cd++].[O-]C([O-])=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1581218
View Documents
Thermo Scientific Chemicals
A1581218
50 g
Each for $53.73
 
AAA1581230
View Documents
Thermo Scientific Chemicals
A1581230
250 g
Each for $142.86
 
AAA158120B
View Documents
Thermo Scientific Chemicals
A158120B
1000 g
Each for $438.27
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

513-78-0
172.42
GKDXQAKPHKQZSC-UHFFFAOYSA-L
10564
CCdO3
MFCD00010918
cadmium carbonate, mikrokalcit, otavite, kalcit, supermikrokalcit, carbonic acid, cadmium salt 1:1, cadmium monocarbonate, chemcarb cdco3, caswell no. 134a, unii-616zvs3bly
[Cd++].[O-]C([O-])=O

Specifications

500°C (decomposition)
50 g
MFCD00010918
14,1616
GKDXQAKPHKQZSC-UHFFFAOYSA-L
cadmium(2+) carbonate
10564
97%
4.26
513-78-0
CCdO3
UN2570
cadmium carbonate, mikrokalcit, otavite, kalcit, supermikrokalcit, carbonic acid, cadmium salt 1:1, cadmium monocarbonate, chemcarb cdco3, caswell no. 134a, unii-616zvs3bly
[Cd++].[O-]C([O-])=O
172.42
172.42
Odorless
Cadmium carbonate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P201-P202-P260-P264b-P270-P271-P281-P284-P301+P312-P302+P352-P304+P340-P308+P313-P310-P312-P330-P363-P501c

H302+H312-H330-H340-H350-H361-H372

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 208-168-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only