missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cadmium nitrate tetrahydrate, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 212342500

 View more versions of this product

Catalog No. AC212342500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

132°C
59°C
10022-68-1
Crystals or Powder
99% min. (Complexometry)
Cd(NO3)2·4H2O
15, 1627
(10% in water) Clear colorless
[Cd++].O[N+]([O-])=O.O[N+]([O-])=O
238.44
CHEBI:86156
≥99%
Cadmium nitrate tetrahydrate
p.a.
Colorless or White
250 g
CdH2N2O6
MFCD00149626
cadmium nitrate tetrahydrate, nitric acid, cadmium salt, tetrahydrate, cd no3 2.4h2o, acmc-20aljb, ksc174c1h, cadmium nitrate-water 1/4, cadmiumnitratetetrahydrate, cadmium ii nitrate tetrahydrate, nitric acid, cadmium salt tetrahydrate
IZIPRBSCGMISKX-UHFFFAOYSA-N
cadmium(2+) bis(nitric acid)
56924536
308.46
Glass bottle

Chemical Identifiers

10022-68-1
238.44
IZIPRBSCGMISKX-UHFFFAOYSA-N
56924536
[Cd++].O[N+]([O-])=O.O[N+]([O-])=O
CdH2N2O6
MFCD00149626
cadmium nitrate tetrahydrate, nitric acid, cadmium salt, tetrahydrate, cd no3 2.4h2o, acmc-20aljb, ksc174c1h, cadmium nitrate-water 1/4, cadmiumnitratetetrahydrate, cadmium ii nitrate tetrahydrate, nitric acid, cadmium salt tetrahydrate
CHEBI:86156
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Harmful in contact with skin.
Fatal if inhaled.
May cause cancer.
May cause genetic defects.
May damage fertility or the unborn child.
Causes damage to organs through prolonge

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/p

GHS Signal Word: Danger

RTECSNumber : EV1850000

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only