Learn More
Cadmium sulfate, anhydrous, ACS, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01186114
Description
Cadmium sulfate is used in pigments and electroplating. It acts as an accelerator in cement formation. It is also employed as lubricants. Further, it is useful for the electroplating of cadmium in electronic circuits. It serves as a precursor to prepare cadmium-based pigment like cadmium sulfide. In the Western standard cell, it is used for electrolyte.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cadmium sulfate | |
For analysis ACS | |
White | |
25 g | |
CdO4S | |
cadmium sulfate, cadmium sulphate, cadmium monosulfate, sulfuric acid, cadmium salt 1:1, cadmium sulfuricum, cadmium sulfate 1:1, cadmium sulfate, tetrahydrate, cadmium 2+ sulfate, caswell no. 136c, unii-947unf3z6o | |
QCUOBSQYDGUHHT-UHFFFAOYSA-L | |
cadmium(2+);sulfate | |
24962 | |
208.46 | |
Hygroscopic | |
4.691 |
1000°C | |
10124-36-4 | |
Powder | |
>99% | |
MFCD00010923 | |
Soluble in water. Slightly soluble in methanol and ethyl acetate. Insoluble in ethanol. | |
[O-]S(=O)(=O)[O-].[Cd+2] | |
208.47 | |
CHEBI:50292 | |
ACS | |
White powder |
Chemical Identifiers
10124-36-4 | |
208.47 | |
QCUOBSQYDGUHHT-UHFFFAOYSA-L | |
24962 | |
cadmium(2+);sulfate |
CdO4S | |
MFCD00010923 | |
cadmium sulfate, cadmium sulphate, cadmium monosulfate, sulfuric acid, cadmium salt 1:1, cadmium sulfuricum, cadmium sulfate 1:1, cadmium sulfate, tetrahydrate, cadmium 2+ sulfate, caswell no. 136c, unii-947unf3z6o | |
CHEBI:50292 | |
[O-]S(=O)(=O)[O-].[Cd+2] |
Safety and Handling
P201-P202-P260-P264b-P270-P271-P280-P301+P310-P304+P340-P308+P313-P310-P330-P501c
H301-H330-H340-H350-H360FD-H372
DOTInformation : DOT Class: 6.1, Packaging Group: III
EINECSNumber : 233-331-6
RTECSNumber : EV2700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only