Learn More
Caffeine, 99.7%
CAS: 58-08-2 | C8H10N4O2 | 194.19 g/mol
$70.45 - $367.18
Identifiants chimiques
| CAS | 58-08-2 |
|---|---|
| Molecular Formula | C8H10N4O2 |
| Molecular Weight (g/mol) | 194.19 |
| MDL Number | MFCD00005758 |
| InChI Key | RYYVLZVUVIJVGH-UHFFFAOYSA-N |
| Synonym | caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep |
| PubChem CID | 2519 |
| ChEBI | CHEBI:27732 |
| IUPAC Name | 1,3,7-trimethylpurine-2,6-dione |
| SMILES | CN1C=NC2=C1C(=O)N(C)C(=O)N2C |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
AA3921414
|
Thermo Scientific Chemicals
03921414 |
25 g |
chaque for $70.45
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
AA3921422
|
Thermo Scientific Chemicals
03921422 |
100 g |
chaque for $118.79
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
AA3921436
|
Thermo Scientific Chemicals
03921436 |
500 g |
chaque for $367.18
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
It acts as a stimulant drug and a reversible acetylcholinesterase inhibitor. It plays an important role for the inactivation of Escherichia coli O 157:H 7 in brain heart infusion (BHI) broth. It is also useful for the protection from photodamage after ultraviolet irradiation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 58-08-2 | |
| 194.19 | |
| RYYVLZVUVIJVGH-UHFFFAOYSA-N | |
| 2519 | |
| 1,3,7-trimethylpurine-2,6-dione |
| C8H10N4O2 | |
| MFCD00005758 | |
| caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep | |
| CHEBI:27732 | |
| CN1C=NC2=C1C(=O)N(C)C(=O)N2C |
Spécifications
| 58-08-2 | |
| 1.23 g/mL | |
| 99.7% | |
| MFCD00005758 | |
| 17705 | |
| Soluble in water,pyridine and ethyl acetate. Partially soluble in acetone,alcohol and petroleum ether. | |
| CN1C=NC2=C1C(=O)N(C)C(=O)N2C | |
| 194.19 | |
| CHEBI:27732 | |
| Powder |
| 234°C to 236°C | |
| 178°C (sublimation) | |
| C8H10N4O2 | |
| 25 g | |
| caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep | |
| RYYVLZVUVIJVGH-UHFFFAOYSA-N | |
| 1,3,7-trimethylpurine-2,6-dione | |
| 2519 | |
| 194.19 | |
| Caffeine |
Sécurité et manipulation
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 200-362-1
missing translation for 'rtecsNumber' : EV6475000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only