missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Caffeine Anhydrous MP Biomedicals

Supplier:  MP Biomedicals Inc 0215011480

 View more versions of this product

Catalog No. ICN15011480


Explore available promotions

Description

Description

Specifications

Specifications

Caffeine Anhydrous
58-08-2
1.23g/mL
MFCD00005758
caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep
CN1C=NC2=C1C(=O)N(C)C(=O)N2C
194.19
CHEBI:27732
Powder
Anhydrous
White
C8H10N4O2
100 g
RYYVLZVUVIJVGH-UHFFFAOYSA-N
1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
2519
194.2

Chemical Identifiers

58-08-2
194.19
RYYVLZVUVIJVGH-UHFFFAOYSA-N
2519
1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
C8H10N4O2
MFCD00005758
caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep
CHEBI:27732
CN1C=NC2=C1C(=O)N(C)C(=O)N2C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at room temperature (15° to 30°C).

SDS
Documents

Documents

Promotions

Promotions