missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Okadaic Acid, Prorocentrum sp.
Okadaic Acid, CAS 78111-17-8, is a highly potent inhibitor of protein phosphatase 1 (IC50 = 10-15 nM) and 2A (IC50 = 0.1 nM).
$1984.76 - $1984.76
Specifications
| Color | Colorless |
|---|---|
| Target Proteins | Protein phosphatase |
| CAS | 78111-17-8 |
| Molecular Formula | C44H68O13 |
| Formula Weight | 805g/mol |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
49-560-4500UG
|
MilliporeSigma™
495604500UG |
500 μg |
Each for $1,984.76
|
|
|||||
Description
Packaged Under Inert GasSpecifications
| Colorless | |
| 78111-17-8 | |
| 805g/mol | |
| Protect from light | |
| 95% by HPLC |
| Protein phosphatase | |
| C44H68O13 | |
| MeOH, EtOH, DMF, CHCl3 | |
| Glassy Solid |