missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Calbiochem™ Aphidicolin
Aphidicolin, CAS 38966-21-1, is a cell-permeable antibiotic that acts as a cell synchronization agent. Blocks the cell cycle at early S-phase.
Supplier: MilliporeSigma™ 1782731MG
Description
Cell PermeableSpecifications
White | |
DNA polymerase α, DNA polymerase δ | |
C20H34O4 | |
DMSO, EtOH, or MeOH | |
1 mg |
98% by TLC | |
38966-21-1 | |
338.5g/mol | |
Solid |