missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Calbiochem™ Clelandâs Reagent, ULTROL™ Grade
Protective agent for SH groups
Supplier: MilliporeSigma™ 2331535GM
Description
- Absorbance: (100mM, 280nm): ≤0.05
- Oxidized dithiothreitol: ≤0.5%
Specifications
154.24 | |
3483-12-3 | |
2°C to 8°C | |
5 g | |
EtOH, Water |
≥99% by titration | |
C4H10O2S2 | |
Solid | |
Dithiothreitol, DTT | |
Calbiochem Clelandâs Reagent |