MilliporeSigma™ Calbiochem™ Complement C3-Depleted Serum, Human
For detection and quantitation of hemolytically active complement components
Supplier: MilliporeSigma™ 2344031ML
Specifications
≤ -70°C | |
1 mL | |
In 150mM NaCl, 50mM sodium phosphate, pH 7.2 |
Liquid | |
Negative for HBsAg and for HCV, HIV-I, and HIV-II antibodies |
For Research Use Only