missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Calbiochem™ H-Gly-Arg-Ala-Asp-Ser-Pro-OH
Inactive control peptide for other fibronectin inhibitors
$130.02 - $397.00
Specifications
Purity | ≥98% by HPLC |
---|---|
Color | White to Off-white |
Content And Storage | -20°C |
Format | Lyophilized Solid |
Formulation | Supplied as a trifluoroacetate salt |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
033400525MG
|
MilliporeSigma™
033400525MG |
5 mg |
Each for $130.02
|
|
|||||
0334005225M
|
MilliporeSigma™
0334005225M |
25 mg |
Each for $397.00
|
|
|||||
Specifications
≥98% by HPLC | |
-20°C | |
Supplied as a trifluoroacetate salt | |
H-Gly-Arg-Ala-Asp-Ser-Pro-OH |
White to Off-white | |
Lyophilized Solid | |
H-Gly-Arg-Ala-Asp-Ser-Pro-OH |
For Research Use Only