missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Calbiochem™ N-Glycolylneuraminic Acid

Formed by action of CMP-Neu5Ac hydroxylase.

Supplier:  MilliporeSigma™ 36200010MG

Catalog No. 36200010MG


Explore available promotions

Description

Description

  • Produced by CMP-Neu5Ac hydroxylase
  • Assay: ≥98% by HPLC
  • CAS: 1113-83-3
  • Molecular Formula: C11H19NO10
  • Color: White
Specifications

Specifications

10 mg
-20°C
SDS
Documents

Documents

Promotions

Promotions