missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Calbiochem™ NG-Hydroxy-L-arginine, Monoacetate Salt

Serves as key intermediate in biosynsis of nitric oxide by constitutive nitric oxide synthase.

Supplier:  MilliporeSigma™ 3992505MG

Catalog No. 3992505MG


Explore available promotions

Description

Description

EMD Millipore Calbiochem™ NG-Hydroxy-L-arginine, Monoacetate Salt is potent inhibitor of liver macrophage arginase.

  • Cell permeable
  • Oxidized to nitric oxide citrulline by cytochrome P450 system
  • CAS: 53598-01-9
  • Molecular formula: C6H14N4O3 · CH3CO2H
  • Color: White
Specifications

Specifications

Purity: ≥98% by Elemental Analysis
−20°C
5 mg
Liver and macrophage arginase
NG-Hydroxy-L-arginine, Monoacetate Salt, Packaged in Glass Bottle
Videos
SDS
Documents

Documents

Promotions

Promotions