missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Calbiochem™ Nω-Hydroxy-nor-L-arginine, Diacetate Salt

Serves as potent, selective, competitive, high affinity inhibitor of arginase.

Supplier:  MilliporeSigma™ 3992755MG

 View more versions of this product

Catalog No. 3992755MG

Explore available promotions

Description

Description

EMD Millipore Calbiochem™ Nω-Hydroxy-nor-L-arginine, Diacetate Salt is shown to specifically interact with manganese-cluster of enzyme active site.

  • Inhibits arginase from rat liver (IC50 = 2μM) mouse macrophages (IC50 = 50μM)
  • Does not function as substrate or inhibitor for NOS
  • Useful tool to study interaction between arginase NOS
  • CAS: 189302-40-7
  • Molecular formula: C5H12N4O3 · 2CH3CO2H
  • Color: Off-White
Specifications

Specifications

Arginase from rat liver
−20°C
Nω-Hydroxy-nor-L-arginine, Diacetate Salt, Packaged in Glass Bottle
5mg
Videos
SDS
Documents

Documents

Promotions

Promotions