missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Calbiochem™ THIOLYTE™ Monobromobimane Reagent, Fluorescent Labeling Compound
Used to label E.coli thioredoxin.
Supplier: MilliporeSigma™ 596105250MG
Description
EMD Millipore Calbiochem™ THIOLYTE Monobromobimane Reagent, Fluorescent Labeling Compound reacts with thiols to yield fluorescent stable thioers.
- Weakly fluorescent
- Reacts with thiols
- Yields highly fluorescent stable thioethers
- Assay: ≥95% by HPLC
- CAS: 71418-44-5
- Packaging: Glass Bottle
- Color: Yellow
Recommended Storage
15° to 30°C
Specifications
250 mg | |
15°C to 30°C | |
THIOLYTE™ MB Reagent, MBB | |
Fluorescent Label |