missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Calbiochem™ Z-Phe-Arg-7-amido-4-methylcoumarin, Hydrochloride
Used as substrate for fluorogenic assay of plasma and glandular kallikreins. MilliporeSigma™; Calbiochem™ Z-Phe-Arg-7-amido-4-methylcoumarin, Hydrochloride also acts as substrate for cathepsin B, cathepsin L, and papain.
Supplier: MilliporeSigma™ 0332150125MG
Description
- Form: White to off-white solid
- Purity: ≥98% by HPLC
Specifications
Solid | |
White to off-white solid | |
Store at -20°C. |
25mg | |
Z-Phe-Arg-7-amido-4-methylcoumarin, Hydrochloride |