Learn More
Calciferol, 97%, Thermo Scientific Chemicals
$64.26 - $257.27
Chemical Identifiers
CAS | 50-14-6 |
---|---|
Molecular Formula | C28H44O |
Molecular Weight (g/mol) | 396.66 |
MDL Number | MFCD00166988,MFCD11656674 |
InChI Key | MECHNRXZTMCUDQ-VLOQVYPSSA-N |
Synonym | ergocalciferol, vitamin d2, calciferol, viosterol, oleovitamin d2, ercalciol, ergorone, detalup, condocaps, crystallina |
PubChem CID | 134129658 |
IUPAC Name | (1S)-3-[2-[(1S,7aR)-1-[(2R,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol |
SMILES | CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1/C[C@@H](O)CCC1=C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6216303
|
Thermo Scientific Chemicals
J6216303 |
1 g |
Each for $64.26
|
|
|||||
AAJ6216306
|
Thermo Scientific Chemicals
J6216306 |
5 g |
Each for $257.27
|
|
|||||
Description
Calciferol has a strong inhibitory effect against bladder tumor promotion by sodium saccharin and induces cell differentiation in leukemia cells. Calciferol has also been shown to be an inhibitor of DNA polymerase. Calciferol with calcium is used to treat or prevent bone loss (osteoporosis). Calciferol is also used with other medications to treat low levels of calcium or phosphate caused by certain disorders (such as hypoparathyroidism, pseudohypoparathyroidism, familial hypophosphatemia). It is often used in kidney disease to keep calcium levels normal and allow normal bone growth.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in ethanol (80 mg/ml at 25°C), water (<1mg/mL at 25°C), methanol, DMF (∼20 mg/ml), and DMSO (20 mg/ml at 25°C).
Notes
Store at 4°C. Incompatible with heat and oxidizing agents.
Chemical Identifiers
50-14-6 | |
396.66 | |
MECHNRXZTMCUDQ-VLOQVYPSSA-N | |
134129658 | |
CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1/C[C@@H](O)CCC1=C |
C28H44O | |
MFCD00166988,MFCD11656674 | |
ergocalciferol, vitamin d2, calciferol, viosterol, oleovitamin d2, ercalciol, ergorone, detalup, condocaps, crystallina | |
(1S)-3-[2-[(1S,7aR)-1-[(2R,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol |
Specifications
50-14-6 | |
White | |
MFCD00166988,MFCD11656674 | |
UN2811 | |
14,10018 | |
Soluble in ethanol (80mg/ml at 25°C),water (<1mg/ml at 25°C),methanol,DMF (∽20mg/ml),and DMSO (20mg/ml at 25°C). | |
+110° (c=1.5 in Ethanol) | |
(1S)-3-[2-[(1S,7aR)-1-[(2R,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol | |
134129658 | |
97% | |
Calciferol |
118°C to 119°C | |
C28H44O | |
1 g | |
1916682 | |
ergocalciferol, vitamin d2, calciferol, viosterol, oleovitamin d2, ercalciol, ergorone, detalup, condocaps, crystallina | |
MECHNRXZTMCUDQ-VLOQVYPSSA-N | |
CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1/C[C@@H](O)CCC1=C | |
396.66 | |
396.66 | |
Powder |
Safety and Handling
GHS H Statement
H301-H311-H330-H372
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P314-P330-P361-P363-P501c
H301+H311-H330-H372
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 200-014-9
RTECSNumber : KE1050000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only