missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium Acetate, EP, USP, 99-100.5%, Spectrum™ Chemical

C1067, 62-54-4, C4H6CaO4

Supplier:  Spectrum Chemical Mfg Cor C1067500GM

 View more versions of this product

Catalog No. 18601203


Explore available promotions

Description

Description

Spectrum™ Chemical Calcium Acetate, USP, EP is used to prevent high blood phosphate levels in patients who are on dialysis due to severe kidney disease. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Specifications

62-54-4
7.2-8.2
C4H6CaO4
VSGNNIFQASZAOI-UHFFFAOYSA-L
calcium diacetate
99 to 100.5%
Amber Glass Bottle
100%
500 g
MFCD00012448
[Ca++].CC([O-])=O.CC([O-])=O
158.17
USP

Chemical Identifiers

62-54-4
158.17
VSGNNIFQASZAOI-UHFFFAOYSA-L
[Ca++].CC([O-])=O.CC([O-])=O
C4H6CaO4
MFCD00012448
calcium diacetate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions