missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcium benzoate hydrate, 98%, water <8%, Thermo Scientific Chemicals
$88.22 - $1160.63
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1235822
|
Thermo Scientific Chemicals
A1235822 |
100 g |
Each for $88.22
|
|
|||||
AAA1235836
|
Thermo Scientific Chemicals
A1235836 |
500 g |
Each for $293.21
|
|
|||||
AAA123580E
|
Thermo Scientific Chemicals
A123580E |
2500 g |
Each for $1,160.63
|
|
|||||
Description
Calcium benzoate hydrate is used in used in the food industry as a preservative.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCalcium benzoate hydrate is used in used in the food industry as a preservative.
Solubility
Soluble in water (2.6 g/100ml at 0°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
2090-05-3 |
Specifications
2090-05-3 | |
MFCD00167164 | |
bis 4-glycidyloxyphenyl methane, bis 4-glycidyloxy phenyl methane, bis 4-oxiran-2-yl methoxy phenyl methane, para-para-bfdge, p,p-bfdge, diphenylolmethane diglycidyl ether, 2,2'-methylenebis p-phenyleneoxymethylene bisoxirane, bis p-2,3-epoxypropoxy phenyl methane, 4,4'-methylenebisphenol diglycidyl ether, oxirane, 2,2'-methylenebis 4,1-phenyleneoxymethylene bis | |
XUCHXOAWJMEFLF-UHFFFAOYNA-N | |
2-[[4-[[4-(oxiran-2-ylmethoxy)phenyl]methyl]phenoxy]methyl]oxirane | |
91511 | |
98% |
C19H20O4 | |
100 g | |
Soluble in water (2.6g/100ml at 0°C). | |
C1C(O1)COC2=CC=C(C=C2)CC3=CC=C(C=C3)OCC4CO4 | |
312.37 | |
282.32 (Anhydrous) | |
Calcium benzoate hydrate, Water <8% |
Safety and Handling
EINECSNumber : 218-235-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only