Learn More
Calcium bromide, anhydrous, 99.5%, Thermo Scientific Chemicals
For Photography, Dehydrating Agent, Manufacturing of Mineral Water
$63.11 - $605.07
Chemical Identifiers
CAS | 7789-41-5 |
---|---|
Molecular Formula | Br2Ca |
Molecular Weight (g/mol) | 199.886 |
MDL Number | MFCD00010902 |
InChI Key | WGEFECGEFUFIQW-UHFFFAOYSA-L |
Synonym | calcium bromide, calcium dibromide, kalziumbromid, brocal, 87cny2eebh, ksc377c0p, calciumbromide, calcium 2+ dibromide, calcium bromide, anhydrous 10g |
PubChem CID | 5360316 |
ChEBI | CHEBI:31338 |
IUPAC Name | calcium;dibromide |
SMILES | [Ca+2].[Br-].[Br-] |
Description
Calcium bromide is a dehydrating agent used in photography, food preservatives, wood preservatives, fire retardants and freezing mixtures. It is used as an oxidizer for mercury emissions control. It is widely used for the preparation of drilling fluids, completion fluids and drilling muds formulations. It also finds application in the manufacture of mineral water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7789-41-5 | |
199.886 | |
WGEFECGEFUFIQW-UHFFFAOYSA-L | |
5360316 | |
calcium;dibromide |
Br2Ca | |
MFCD00010902 | |
calcium bromide, calcium dibromide, kalziumbromid, brocal, 87cny2eebh, ksc377c0p, calciumbromide, calcium 2+ dibromide, calcium bromide, anhydrous 10g | |
CHEBI:31338 | |
[Ca+2].[Br-].[Br-] |
Specifications
806°C to 812°C | |
7789-41-5 | |
Powder | |
99.5% | |
MFCD00010902 | |
Soluble in water,methanol,ethanol and acetone. Insoluble in dioxane,chloroform and ether. | |
[Ca+2].[Br-].[Br-] | |
199.886 | |
CHEBI:31338 | |
Hygroscopic | |
3.353 |
730°C | |
White | |
10 g | |
Br2Ca | |
calcium bromide, calcium dibromide, kalziumbromid, brocal, 87cny2eebh, ksc377c0p, calciumbromide, calcium 2+ dibromide, calcium bromide, anhydrous 10g | |
WGEFECGEFUFIQW-UHFFFAOYSA-L | |
calcium;dibromide | |
5360316 | |
199.9 | |
White to off-white powder or beads | |
Calcium bromide, Anhydrous |
Safety and Handling
P280-P305+P351+P338-P310
H318
EINECSNumber : 232-164-6
RTECSNumber : EV9328000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only