missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium carbide, 72-82%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 389790010

 View more versions of this product

Catalog No. AC389790010

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2300.0°C
2160.0°C
75-20-7
1 kg
C2Ca
MFCD00010905
calcium carbide, 3-phenoxypropylene di acetate, glycerol phenyl ether diacetate, 3-phenoxy-1,2-propanediol diacetate, phenylglyceryl ether diacetate, 1-phenyl ether-2,3-diacetate glycerol, 1,2-propanediol, 3-phenoxy-, diacetate, 1-acetyloxy-3-phenoxypropan-2-yl acetate, quartz crystal, 3-phenoxypropane-1,2-diyl diacetate
64.1
72 to 82%
Glass bottle
Calcium carbide
Gray
Lump
12 to 23% (CaO) (Complexometry)
CaC2
15, 1658
Solubility in water: hydrolyses
64.1
Positive

Chemical Identifiers

75-20-7
64.1
calcium carbide, 3-phenoxypropylene di acetate, glycerol phenyl ether diacetate, 3-phenoxy-1,2-propanediol diacetate, phenylglyceryl ether diacetate, 1-phenyl ether-2,3-diacetate glycerol, 1,2-propanediol, 3-phenoxy-, diacetate, 1-acetyloxy-3-phenoxypropan-2-yl acetate, quartz crystal, 3-phenoxypropane-1,2-diyl diacetate
C2Ca
MFCD00010905
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
In contact with water releases flammable gases which may ignite spontaneously.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.

GHS P Statement
Keep away from any possible contact with water,because of violent reaction and possible flash fire.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protecti

GHS Signal Word: Danger

EINECSNumber : 200-848-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only