missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcium Carbonate (Powder/USP/FCC), Fisher Chemical™
Supplier: Thermo Fisher Scientific FLC6310
Specifications
Calcium Carbonate | |
471-34-1 | |
8 to 9 | |
10 kg | |
MFCD00010906 | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
calcium carbonate | |
10112 | |
100.09 | |
FCC/USP | |
2.0% max. (FCC), 2.0% max. (FCC/USP), 2.0% max. (USP) |
825°C | |
White | |
Solid | |
CCaO3 | |
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
[Ca++].[O-]C([O-])=O | |
100.09 | |
CHEBI:3311 | |
98.0 to 100.5% (FCC) | |
Pass Test (FCC), Pass Test (FCC/USP), Pass Test (USP) | |
Fiber Drum |
Chemical Identifiers
471-34-1 | |
100.09 | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
10112 | |
calcium carbonate |
CCaO3 | |
MFCD00010906 | |
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
CHEBI:3311 | |
[Ca++].[O-]C([O-])=O |
Safety and Handling
WARNING!
Emergency Overview
Irritating to eyes and skin. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:2
Flammability:0
Instability:0