missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcium Chloride, Anhydrous (Pellets, 4-20 mesh, for Desiccators), Fisher Chemical™
$505.38 - $1187.31
Chemical Identifiers
CAS | 10043-52-4 |
---|---|
Molecular Formula | CaCl2 |
Molecular Weight (g/mol) | 110.98 |
MDL Number | MFCD00010903 |
InChI Key | UXVMQQNJUSDDNG-UHFFFAOYSA-L |
PubChem CID | 5284359 |
ChEBI | CHEBI:3312 |
IUPAC Name | calcium dichloride |
SMILES | [Cl-].[Cl-].[Ca++] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
C614-500
|
Thermo Fisher Scientific
FLC614500 |
500 g | Poly Bottle |
Each for $517.23
Case of 6 Each for $3,103.38
|
|
||||
C614-3
|
Thermo Fisher Scientific
FLC6143 |
3 kg | Poly Bottle |
Each for $505.38
Case of 4 Each for $2,021.52
|
|
||||
C614-10
|
Thermo Fisher Scientific
FLC61410 |
10 kg | Poly Pail |
Each for $1,187.31
|
|
||||
Chemical Identifiers
10043-52-4 | |
110.98 | |
UXVMQQNJUSDDNG-UHFFFAOYSA-L | |
CHEBI:3312 | |
[Cl-].[Cl-].[Ca++] |
CaCl2 | |
MFCD00010903 | |
5284359 | |
calcium dichloride |
Specifications
782°C | |
Solid | |
Beige | |
2.15g/mL | |
500 g | |
CaCl2 | |
4 to 20 Mesh | |
UXVMQQNJUSDDNG-UHFFFAOYSA-L | |
calcium dichloride | |
5284359 | |
110.99 |
1600°C | |
8 to 10 | |
10043-52-4 | |
Calcium Chloride | |
Pass Test | |
MFCD00010903 | |
Poly Bottle | |
[Cl-].[Cl-].[Ca++] | |
110.98 | |
CHEBI:3312 | |
Calcium Chloride Anhydrous, For desiccators |
Safety and Handling
WARNING!
Emergency Overview
Irritating to eyes. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.
NFPA
Health:2
Flammability:0
Instability:1