missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium citrate tribasic tetrahydrate, 98+%, for analysis
CAS: 5785-44-4 | C12H10Ca3O14·4H2O | 570.5 g/mol
$64.72 - $157.03
Chemical Identifiers
| CAS | 5785-44-4 |
|---|---|
| Molecular Formula | C12H10Ca3O14·4H2O |
| Molecular Weight (g/mol) | 570.5 |
| MDL Number | MFCD00150786 |
| InChI Key | LNIZKKFWMDARJV-UHFFFAOYSA-H |
| Synonym | calcium citrate tetrahydrate, unii-mlm29u2x85, calcium citrate usp, tricalcium dicitrate tetrahydrate, citric acid calcium salt, citracal tn, calcium citrate 3:2 , tetrahydrate, acmc-20aliq, ksc493e4f |
| PubChem CID | 5282392 |
| IUPAC Name | tricalcium;2-hydroxypropane-1,2,3-tricarboxylate;tetrahydrate |
| SMILES | C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.O.[Ca+2].[Ca+2].[Ca+2] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC390082500
|
Thermo Scientific Chemicals
390082500 |
250 g | Plastic bottle |
Each for $64.72
|
|
||||
|
AC390080010
|
Thermo Scientific Chemicals
390080010 |
1 kg | Plastic bottle |
Each for $157.03
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 5785-44-4 | |
| 570.5 | |
| LNIZKKFWMDARJV-UHFFFAOYSA-H | |
| 5282392 | |
| C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.O.[Ca+2].[Ca+2].[Ca+2] |
| C12H10Ca3O14·4H2O | |
| MFCD00150786 | |
| calcium citrate tetrahydrate, unii-mlm29u2x85, calcium citrate usp, tricalcium dicitrate tetrahydrate, citric acid calcium salt, citracal tn, calcium citrate 3:2 , tetrahydrate, acmc-20aliq, ksc493e4f | |
| tricalcium;2-hydroxypropane-1,2,3-tricarboxylate;tetrahydrate |
Specifications
| 5785-44-4 | |
| Authentic | |
| Plastic bottle | |
| MFCD00150786 | |
| 15, 1663 | |
| Solubility in water: 1g/L (20°C) | |
| C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.O.[Ca+2].[Ca+2].[Ca+2] | |
| 570.5 | |
| 570.5 | |
| Analysis | |
| Liquid |
| 10.0 % to 13.3 % (150°C, 4 hrs) | |
| 98.0 to 102% (on dried basis) (Complexometry) | |
| C12H10Ca3O14·4H2O | |
| 250 g | |
| calcium citrate tetrahydrate, unii-mlm29u2x85, calcium citrate usp, tricalcium dicitrate tetrahydrate, citric acid calcium salt, citracal tn, calcium citrate 3:2 , tetrahydrate, acmc-20aliq, ksc493e4f | |
| LNIZKKFWMDARJV-UHFFFAOYSA-H | |
| tricalcium;2-hydroxypropane-1,2,3-tricarboxylate;tetrahydrate | |
| 5282392 | |
| 98+% | |
| 0.2% max. (in HCl) | |
| Calcium citrate tribasic tetrahydrate |
RUO – Research Use Only