Learn More
Calcium crystalline dendritic pieces, 99.98% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01445714
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Calcium crystalline dendritic pieces | |
(metals basis) | |
MFCD00010897 MFCD00085314 | |
25 g | |
OYPRJOBELJOOCE-UHFFFAOYSA-N | |
calcium | |
5460341 | |
99.98% | |
Ampule |
7440-70-2 | |
Ca | |
UN1401 | |
unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate | |
[Ca] | |
40.08 | |
CHEBI:29320 | |
Odorless | |
Pieces |
Chemical Identifiers
7440-70-2 | |
40.08 | |
OYPRJOBELJOOCE-UHFFFAOYSA-N | |
5460341 | |
calcium |
Ca | |
MFCD00010897 MFCD00085314 | |
unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate | |
CHEBI:29320 | |
[Ca] |
Safety and Handling
GHS H Statement
H261
In contact with water releases flammable gas.
P223-P231+P232-P280-P335+P334-P370+P378q-P501c
H261
DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: CALCIUM
EINECSNumber : 213-179-5
RTECSNumber : EV8040000
TSCA : Yes
storageNote1 :
- Ampouled under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only