missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium cyanamide, tech., Thermo Scientific Chemicals

$89.32 - $284.89

Chemical Identifiers

CAS 156-62-7
Molecular Formula CCaN2
MDL Number MFCD00064894
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA8946222
View Documents
Thermo Scientific Chemicals
08946222
100 g
Each for $89.32
 
AA8946236
View Documents
Thermo Scientific Chemicals
08946236
500 g
Each for $284.89
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

156-62-7
CCaN2
MFCD00064894

Specifications

1340°C
Gray
100 g
CaNCN
UN1403
14,1662
Technical
Moisture sensitive
Calcium cyanamide
156-62-7
Powder
CCaN2
MFCD00064894
4124391
80.11
Specific type
2.29 g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H335
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P304+P340-P305+P351+P338-P310-P312-P330-P501c

H302-H318-H335

DOTInformation : Transport Hazard Class: 4.3; Packing Group: III; Proper Shipping Name: CALCIUM CYANAMIDE

EINECSNumber : 205-861-8

RTECSNumber : GS6000000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only