Learn More
Calcium dihydrogen phosphate hydrate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1212336
Description
Calcium dihydrogen phosphate hydrate is a pH control reagent, stabilizer for plastics, manufacture of glass.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCalcium dihydrogen phosphate hydrate is a pH control reagent, stabilizer for plastics, manufacture of glass.
Solubility
Soluble in water 18 g/L @ 30°C.
Notes
Loses H{2}O at 100°, dec. at 200°
Specifications
Calcium dihydrogen phosphate hydrate | |
301524-28-7 | |
CaH4O8P2 | |
None | |
Calcium phosphate, monobasic; Calcium tetrahydrogen diphosphate | |
YYRMJZQKEFZXMX-UHFFFAOYSA-L | |
calcium didihydrogen phosphate | |
234.06 (Anhydrous) |
∼200°C (decomposition) | |
500 g | |
MFCD00010898 | |
14,1693 | |
Soluble in water 18g/L @ 30°C. | |
[Ca++].OP(O)([O-])=O.OP(O)([O-])=O | |
234.05 | |
97% |
Chemical Identifiers
301524-28-7 | |
234.05 | |
YYRMJZQKEFZXMX-UHFFFAOYSA-L | |
calcium didihydrogen phosphate |
CaH4O8P2 | |
MFCD00010898 | |
Calcium phosphate, monobasic; Calcium tetrahydrogen diphosphate | |
[Ca++].OP(O)([O-])=O.OP(O)([O-])=O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 231-837-1
RTECSNumber : TB8527000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only